Imprimer
Lysing kits for Fastprep®-24 5G, Lysing Matrix SS, MP Biomedicals
Fournisseur: MP Biomedicals
116540431 116921100
ICNA116540431EA 181 EUR
ICNA116540431 ICNA116921100
Lysing kits for Fastprep®-24 5G, Lysing Matrix SS, MP Biomedicals
Homogénéisateurs Homogénéisateurs de paillasse
Each impact-resistant 2 ml tube contains one 6,35 mm, 0,25 cal stainless-steel grinding ball.
  • Can be stored at room temperature
  • DNase and RNase free
Use to break hard/brittle samples. Specialty applications include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR