Imprimer

Human recombinant Stem Cell Factor (from E. coli), MP Biomedicals

Fournisseur: MP Biomedicals

0215869802
ICNA0215869802EA 87.9 EUR
ICNA0215869802
Human recombinant Stem Cell Factor (from E. coli), MP Biomedicals
Protéines et Peptides

Stem Cell Factor (SCF) is a hematopoietic growth factor that exerts its activity at the early stages of hematopoiesis. It stimulates the proliferation of myeloid, erythroid, and lymphoid progenitors in bone marrow cultures and has been shown to act synergistically with colony stimulating factors.


  • Soluble in water and most aqueous buffers
  • Specific activity: ≤20 ng/ml

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR