Imprimer
Macerozyme, MP Biomedicals
Fournisseur: MP Biomedicals
0215234010
ICNA0215234010EA 186 EUR
ICNA0215234010
Macerozyme, MP Biomedicals
Enzymes
The amount of macerating activity on potato tuber disks as determined by weight loss at 40 °C, pH 5,0.
  • Readily soluble in water.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR