Imprimer

Pyridoxal-5'-phosphate monohydrate ~98%, powder_off-white

Fournisseur: MP Biomedicals

Synonyms: Pyridoxal-5-phosphate hydrate, Pyridoxal 5-phosphate monohydrate, Pyridoxal 5'-phosphate monohydrate, (4-Formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate hydrate

0210276925 0210276901 0210276905
ICNA0210276925EA 456 EUR
ICNA0210276925 ICNA0210276901 ICNA0210276905
Pyridoxal-5'-phosphate monohydrate ~98%, powder_off-white
Pyridoxal-5'-phosphate monohydrate

Pyridoxal-5-Phosphate Monohydrate is an active form of Vitamin B6, an enzyme prosthetic group.


Pyridoxal-5-Phosphate Monohydrate is useful for modifying lysyl and valyl residues in proteins. It acts as a coenzyme in transamination, decarboxylation and deamination reactions such as the conversion of dopa into dopamine; of glutamate into GABA; of histidine to histamine. PLP works mechanistically through formation of a Schiff-base.


Soluble in 1 M Hydrochloric acid (50 mg/ml - clear, colourless to light yellow-green solution).

Formule: C₈H₁₀NO₆P·1H₂O
MW: 265.17 g/mol (247.15 anhy)
Température de stockage: Congélateur
Numéro MDL: MFCD00149414
Numéro CAS: 41468-25-1

Commander maintenant

Résultats des tests de spécification

Identity Test Passes
Purity ~98%
FTIR Conforms to Standard
Moisture Content ~6.8%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR