Imprimer
Ice Cubers, Self Contained, IM
Fournisseur: HOSHIZAKI
IM-100CNE-HC23 IM-100CNE-HC
HOSHIM-100CNE-HC23EA 4130 EUR
HOSHIM-100CNE-HC23 HOSHIM-100CNE-HC
Ice Cubers, Self Contained, IM
Machines à glace
Self contained cube ice makers producing high-quality cube ice. Meeting even the strictest hygiene requirements, this true allrounder offers many application possibilities in segments including the foodservice and medical sector.
  • Uses refrigerant R290, this natural refrigerant is not only climate-friendly, but also boosts the machine, when it comes to performance and production capacity
  • The exterior is made of stainless steel
  • The closed water circuit provides maximum protection against any type of contamination during the entire cubing process
  • Magnetic
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR