Imprimer
Avantor® Hichrom, Female to Female Luer Adapter
Fournisseur: Avantor
HICHHI-314EA 34.6 EUR
HICHHI-314
Avantor® Hichrom, Female to Female Luer Adapter
Raccords et connecteurs
This adapter provides us the opportunity to adapt female luer to female luer connection to a threaded connection.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR