Imprimer
Desalting-RP HPLC columns, AdvanceBio
Fournisseur: Agilent
PL1612-1102
HEWLPL1612-1102EA 582 EUR
HEWLPL1612-1102
Desalting-RP HPLC columns, AdvanceBio
Colonnes pour chromatographie
The key features and benefits of the AdvanceBio Desalting RP columns.
  • Allow MS detection with traditionally non-MS friendly techniques such as IEX, SEC, and affinity chromatography
  • Save time by desalting your samples on-line
  • Concentrate samples prior to elution
  • Reduce down time and maintenance by keeping your MS cleaner
  • Improve overall data confidence by improving the quality of your data
Reversed-phase desalting for on-line removal of salt ions before MS detection affinity, ion exchange, and size exclusion chromatography are commonly used techniquesMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR