Imprimer
Sampling swab, Gynobrush® Plus
Fournisseur: HERENZ HEINZ
1032929
HERE1032929EA 149 EUR
HERE1032929
Sampling swab, Gynobrush® Plus
Écouvillons Écouvillons d'échantillonnage
This Gynobrush® Plus has a soft plastic tip.
Suitable for sampling gynocological fluids.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR