Imprimer
Filtering flasks with glass hose connection
Fournisseur: GLASWARENFABRIK KARL HECHT
41270036 41270042 41270048 41270054
HECH41270036EA 68.5 EUR
HECH41270036 HECH41270042 HECH41270048 HECH41270054
Filtering flasks with glass hose connection
Ballons Fioles à vide
Flasks for vacuum use, made of DURAN® borosilicate glass.
  • Erlenmeyer shape
  • Olive hose connection Ø 11 mm
  • Does not conform to the 'Equipment and product safety regulations'
ISO 6556
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR