Imprimer
Enzyme test pipette, conformity certified, Assistent
Fournisseur: GLASWARENFABRIK KARL HECHT
40545724
HECH40545724EA 124 EUR
HECH40545724
Enzyme test pipette, conformity certified, Assistent
Pipettes Pipettes pour hémomètres
Clear glass, with elongated strong tip, precision type, for partial delivery (not calibrated to tip).
  • Calibrated to deliver (Ex), short delivery time
  • Zero point at top, main points
  • Fused amber graduation 'ASSISTENT-PERMANENT'
Available with individual certificate on request (extra charge).
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR