Imprimer
Test sieves with stainless steel frame
Fournisseur: NEXOPART
206146571 206143990 206150929 206151384 206151582 206152602 206142009 206160973 206153685 145147-01 145325-01 206174413 206164957 206142542 206143259 206165343 206165176 206144515 206168702 206168740 206168795 206134158 206134592 206168917 206173973 206174239 206146953 206166074 206147011 206168986 206168771 206163127 206141415 206141309 206142382 206143082 206144638 206144874 206144966 206146366 206146489 206165626 206167200 206167989 206136350 206136381 206140845 206141330 206141453 206142566 206143426 206144416 206144829 206145000 206145109 206145437 206146007 206149916 206161178 206164667 206168924 206164469 206160959 206165220
HAVE206146571EA 138 EUR
HAVE206146571 HAVE206143990 HAVE206150929 HAVE206151384 HAVE206151582 HAVE206152602 HAVE206142009 HAVE206160973 HAVE206153685 HAVE145147-01 HAVE145325-01 HAVE206174413 HAVE206164957 HAVE206142542 HAVE206143259 HAVE206165343 HAVE206165176 HAVE206144515 HAVE206168702 HAVE206168740 HAVE206168795 HAVE206134158 HAVE206134592 HAVE206168917 HAVE206173973 HAVE206174239 HAVE206146953 HAVE206166074 HAVE206147011 HAVE206168986 HAVE206168771 HAVE206163127 HAVE206141415 HAVE206141309 HAVE206142382 HAVE206143082 HAVE206144638 HAVE206144874 HAVE206144966 HAVE206146366 HAVE206146489 HAVE206165626 HAVE206167200 HAVE206167989 HAVE206136350 HAVE206136381 HAVE206140845 HAVE206141330 HAVE206141453 HAVE206142566 HAVE206143426 HAVE206144416 HAVE206144829 HAVE206145000 HAVE206145109 HAVE206145437 HAVE206146007 HAVE206149916 HAVE206161178 HAVE206164667 HAVE206168924 HAVE206164469 HAVE206160959 HAVE206165220
Test sieves with stainless steel frame
Tamis Tamis analytiques
These test sieves are suitable both for hand sieving as for dry and wet sieving with all Haver test sieve shakers. Bulk material of different branches can be classified and separated with these test sieves; industrial rocks and minerals, food, glass, ceramic, granulates, powders, fertilizer, etc.
  • Smooth surface of frames prevents cross-contamination
  • Sieve fabric retains its tension very well even after intensive use
  • Compatibility with equivalent test sieves manufactured in accordance with current standards
Stainless steel frame with woven wire clothMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR