Imprimer
Adjustable multi-channel gel loading syringe
Fournisseur: HAMILTON BONADUZ
84611
HAMI84611EA 2680 EUR
HAMI84611
Adjustable multi-channel gel loading syringe
Seringues Seringues de chromatographie
Adjustable multi-channel gel loading syringes simplify and speed sample loading of 96 lane gels. The adjustable gel loading syringe (AGLS) allows quick, easy transfer and loading of DNA samples. Eight samples at a time can be transferred directly from a microwell plate with 9 mm spacing to a 96-lane gel with 10,8 mm spacing.
  • Compatible with 96-lane ABI 377 PRISM® DNA sequencing instrument
  • Easy-to-use design for needle spacing adjustment
Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision machined PTFE plunger tipMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR