Imprimer
Hardness quality control standard
Fournisseur: Hach
2833349 2833449
HACH2833349EA 96.6 EUR
HACH2833349 HACH2833449
Hardness quality control standard
Étalons Étalons pour l'analyse environnementale
These mixed standards are ready to use with no dilutions required. Includes standards for both total hardness and calcium hardness determination.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR