Imprimer
Ammonia (nitrogen) test strips
Fournisseur: Hach
2755325
HACH2755325EA 21.2 EUR
HACH2755325
Ammonia (nitrogen) test strips
Kits de tests environnementaux
Ammonia (Nitrogen) is a product of microbiological decay of plant and animal protein. Its presence in raw surface waters usually indicates domestic or agricultural pollution. Above certain levels, it is toxic to fish.
  • Easy to use, disposable, and inexpensive
  • A great way to obtain quick, quantitative answers in the field or in the lab
  • Parameter: ammonia, nitrogen, low range - As NH₃-N, for freshwater
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR