Imprimer

Ferroïne (Phénanthroline-1,10-fer(II) sulfate) solution_SC Indicator, COD_determination

Fournisseur: Hach

Synonyms: Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate

181233
HACH181233EA 25 EUR
HACH181233
Ferroïne (Phénanthroline-1,10-fer(II) sulfate) solution_SC Indicator, COD_determination
Ferroïne (Phénanthroline-1,10-fer(II) sulfate)

Oxidation-reduction indicator solution used in determination of COD by ferrous titration.

Formule: C₃₆H₂₄FeN₆SO₄
MW: 692.54 g/mol
Densité: 0.9999
Numéro MDL: MFCD00036428
Numéro CAS: 14634-91-4

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR