Imprimer

pH Buffer solutions

Fournisseur: Hach

1222149 1222249 1222349 S11M009 1222220 S11M010 S11M011
HACH1222149EA 21.6 EUR
HACH1222149 HACH1222249 HACH1222349 HACHS11M009 HACH1222220 HACHS11M010 HACHS11M011
pH Buffer solutions
Tampons Tampons pH

For calibration of pH meters.


Accuracy: pH is ±0,02 at 25 °C


Certifications: Traceable to NIST.


Attention: Please note that the pH 4 and 10 buffers contain formaldehyde <0,1% and the pH 7 buffer contains formaldehyde 0,1 to 1%.

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR