Imprimer
Real Time RT-PCR Kit
Fournisseur: GOLD STANDARD DIAGNOSTICS
PCOV6111
GSDCPCOV6111EA 700 EUR
GSDCPCOV6111
Real Time RT-PCR Kit
Tests Tests qPCR
High quality RT-PCR kits for detection of various infectious diseases.
  • Fast and reliable results in approximately 1 hour
  • Validated for a variety of specimen types
  • Excellent performance with high sensitivity and specificity
  • Extraction/inhibition and positive control included
  • Suitable for most PCR platforms
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR