Imprimer
1536-well microplates, cyclo olefin
Fournisseur: Greiner Bio-One
792870-906
GREI792870-906EA 1970 EUR
GREI792870-906
1536-well microplates, cyclo olefin
Microplaques Microplaques HTS
Clear, 1536-well cycloolefin storage plates (with optimised geometry for low evaporation) for acoustic liquid handling.
  • Free from DNase, RNase, and human DNA
  • Groove around the plate edge matches cycloolefin plate lid (not included) - designed to reduce evaporation and minimise edge effects
Acoustic liquid dispensing offers the advantage of pipetting small volumes very precisely and with minimal disturbance without the risk of cross-contamination. The process involves the transferral of DMSO-containing solutions in the nanoliter to picoliter range fromMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR