Imprimer
PCR plate centrifuge/vortex mixer, CVP-2
Fournisseur: GRANT INSTRUMENTS
CVP-2
GRANCVP-2EA 1570 EUR
GRANCVP-2
PCR plate centrifuge/vortex mixer, CVP-2
Centrifugeuses Microcentrifugeuses
This unique product offers 3 devices in one: Centrifuge with vortex mixing, PCR plate centrifuge and PCR plate mixer. It allows for the simultaneous sample preparation of multiple samples at one time by providing repeating spin down and shaking of two un, semi or fully skirted PCR plates.
  • Centrifuge and vortex mixer combined for significant time saving
  • Spin and vortex mixing functions can operate independently
  • Very small footprint – saves bench space
  • Simple push-buttons and a clear 2-line LCD status display of set and actual values
  • Clear window in the lid
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR