Imprimer
Corning® Gosselin™ Petri Dish, Aseptic
Fournisseur: Corning
SH91-101
GOSSSH91-101EA 186 EUR
GOSSSH91-101
Corning® Gosselin™ Petri Dish, Aseptic
Boîtes de culture Boîtes de Petri
Transparent PS Petri dishes for food industry laboratories.
  • Diam. 90 mm
  • Crystal Polystyrene (PS)
  • Optimal flatness
  • Easy opening sleeves
  • 1 vent
  • Aseptic
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR