Imprimer
GOJO® Antimicrobial Plus Foam Handwash Refills
Fournisseur: GOJO Industries - Europe Ltd
5348-02-EEU00 5148-03-EEU00
GOJO5348-02-EEU00EA 60.4 EUR
GOJO5348-02-EEU00 GOJO5148-03-EEU00
GOJO® Antimicrobial Plus Foam Handwash Refills
Soin de la peau Savons et soins nettoyants
La mousse lavante antimicrobienne GOJO® sans quat est douce pour la peau et formulée pour être très efficace.
  • Suitable for sensitive skin and formulated for high frequency use
  • Enriched with a moisturising plant-based agent
  • Broad Spectrum efficacy (bacteria, yeasts, and virus)
  • Passes EN 1499 in 30 seconds
  • Targets food specific pathogens such as Salmonella, Listeria, and E. coli
  • Unfragranced and colourless
  • GOJO CLEAN SEALED™ refills snap quickly and easily into place; protects users from the contamination risks of bulk soap
  • Refills made with 100% R-PET
  • Halal
More Product Information
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR