Imprimer
Tubing
Fournisseur: GILSON
F117740 F117742 F117744 F117748 F117749 F117750 F117933 F117934 F117936 F117938 F117940 F117942 F117943 F117945 F117946 F117948 F117949 F117952 F117953 F117954 F117956 F117958 F117960 F117962 F117963 F117965 F1825122 F1817741 F1817743 F1817745 F117966 F117968 F117969 F117970 F117975 F117976 F117977 F117978 F117979 F117980 F117981 F117982 F1825103 F1825111 F1825112 F1825113 F1825114 F1825101 F1825131 F1825132 F1825133 F117935 F117939 F117941 F1825102
GILSF117740EA 100 EUR
GILSF117740 GILSF117742 GILSF117744 GILSF117748 GILSF117749 GILSF117750 GILSF117933 GILSF117934 GILSF117936 GILSF117938 GILSF117940 GILSF117942 GILSF117943 GILSF117945 GILSF117946 GILSF117948 GILSF117949 GILSF117952 GILSF117953 GILSF117954 GILSF117956 GILSF117958 GILSF117960 GILSF117962 GILSF117963 GILSF117965 GILSF1825122 GILSF1817741 GILSF1817743 GILSF1817745 GILSF117966 GILSF117968 GILSF117969 GILSF117970 GILSF117975 GILSF117976 GILSF117977 GILSF117978 GILSF117979 GILSF117980 GILSF117981 GILSF117982 GILSF1825103 GILSF1825111 GILSF1825112 GILSF1825113 GILSF1825114 GILSF1825101 GILSF1825131 GILSF1825132 GILSF1825133 GILSF117935 GILSF117939 GILSF117941 GILSF1825102
Tubing
Tuyaux Transfer Tubing
As part of the MINIPULS® 3 and Evolution lines of peristaltic pumps, Gilson offers a variety of tubing of different material, inner diameter sizes, and lengths.
Silicone (Versilic®) tubing is biologically neutral and does not inhibit cell culture while meeting European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
ISO-VERSINIC® tubing shows excellent resistance to strong acids, oxidising agents, and aromatic and chlorinated solvents.
PharMed® tubing meets European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993More Product Information
Plus About VWR