Imprimer
Restriction enzymes, BpiI, FastDigest™, fermentas
Fournisseur: Thermo Fisher Scientific
FD1014
FD1014EA 45.83 EUR
FD1014
Restriction enzymes, BpiI, FastDigest™, fermentas
Réactifs pour acides nucléiques Restriction Enzymes
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
  • 100% activity of all FastDigest™ enzymes in one universal reaction buffer
  • Designed to complete digestion in 5 to 15 minutes
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR