Imprimer
Aqueous mounting medium, Fluoroshield™ with 1,4-Diazabicyclo[2.2.2]octane, Sigma-Aldrich®
Fournisseur: SIGMA ALDRICH MICROSCOPY
F6937-20ML
F6937-20MLEA 107 EUR
F6937-20ML
Aqueous mounting medium, Fluoroshield™ with 1,4-Diazabicyclo[2.2.2]octane, Sigma-Aldrich®
Colorants Réactifs pour la microscopie et la coloration
Fluoroshield™ with 1,4-Diazabicyclo[2.2.2]octane is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
  • Storage temperature: 2 - 8 °C
Fluoroshield™ prevents rapid photobleaching of FITC, Texas Red®, AMCA, Alexa Fluor™ 488, Alexa Fluor 594, Cy® dyes, and tetramethylrhodamine. This mounting medium is fortified with 1,4-Diazabicyclo[2.2.2]octane (a generic version of DABCO™), an anti-fade reagent used to retard photobleaching of fluorescein and other fluorescent dyes.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR