Imprimer
Conductivity meters, handheld, Eutech CyberScan COND 610
Fournisseur: EUTECH
ECCONWP61043K
EUTEECCONWP61043KEA 1770 EUR
EUTEECCONWP61043K
Conductivity meters, handheld, Eutech CyberScan COND 610
Compteurs électrochimiques Conductimètres
The CyberScan COND 600 series with auto-ranging capabilities measure temperature (with ATC), conductivity, TDS, salinity and resistivity.
  • Mega-memory space that stores/recalls up to 500 data sets with temperature readings, as well as time and date-stamping formatted to meet GLP standards
  • Wireless and cable-free data transfer with infrared IrDA technology
  • Comes with PC-based complimentary Data Acquisition Software for hassle-free data import and report-generation
  • Large, easy-to-read custom screen with simultaneous information display allows optimum viewing
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR