Imprimer
S@feMate ECO+ Microbiological Safety Cabinets, Class II
Fournisseur: BioAir S.p.A.
LDQ320N LDQ420N LDQ520N LDQ620N BI00LDQ6201 BI00LDQ6200 BI00LDQ5201 BI00LDQ5200 BI00LDQ4201 BI00LDQ4200 BI00LDQ3201 BI00LDQ3200
EUCLLDQ320NEA 0 EUR
EUCLLDQ320N EUCLLDQ420N EUCLLDQ520N EUCLLDQ620N EUCLBI00LDQ6201 EUCLBI00LDQ6200 EUCLBI00LDQ5201 EUCLBI00LDQ5200 EUCLBI00LDQ4201 EUCLBI00LDQ4200 EUCLBI00LDQ3201 EUCLBI00LDQ3200
S@feMate ECO+ Microbiological Safety Cabinets, Class II
Hottes et coffrets Postes de sécurité biologique
Safemate ECO+ Cabinets are supplied in four different sizes (0.9, 1.2, 1.5 and 1.8). These last generation Microbiological Safety Cabinets Class II type A2, have been certified according to the most stringent safety standards (EN 12469-2000). The internal design, the air flow aerodynamics and monitoring, the built-in safety devices and the very accurate manufacturing, guarantees the highest performances at the most stringent safety levels, as specified by EN 12469 standard and have been certified by the most prestigious European certification bodies for Safety Cabinets.
Plus About VWR