Imprimer
Precision scale magnifiers
Fournisseur: ESCHENBACH OPTIK
155074
ESCH155074EA 193 EUR
ESCH155074
Precision scale magnifiers
Loupes
Precision magnifiers with brightness control and removable measuring scale available with battery or mains handle with brightness control.
Aspheric PXM® lightweight lens with Cera-tec® hard coating (both sides).
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR