Imprimer
Dust covers
Fournisseur: EP SCIENTIFIC
DC-VOA
EPSCDC-VOAEA 184 EUR
EPSCDC-VOA
Dust covers
Capuchons Bouchons pour flacons de chromatographie
These PE dust covers are designed to fit all standard size VOA vial caps. They prevent contamination and protect critical samples during transport.
  • Allow for septum expansion
  • Pre cleaned dust covers available upon request
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR