Imprimer
ACTH ELISA kit, grow cells
Fournisseur: ENZO LIFE SCIENCES
ENZKIT1380001
ENZOENZKIT1380001EA 730 EUR
ENZOENZKIT1380001
ACTH ELISA kit, grow cells
Tests Tests ELISA
Highly sensitive ACTH ELISA kit enabling detection of ACTH in human plasma in 4.5 hours.
  • Sensitive measurement of ACTH, detecting as little as 0,46 pg/ml.
  • Negligible reactivity to similar hormones.
  • High throughput with results in 4.5 hours.
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of ACTH in plasma in human samples.
The ACTH ELISA kit is a colorimetric immunoassay kit with results in 4,5 hours.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR