Imprimer
TSH ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ENZKIT1310001
ENZOENZKIT1310001EA 382 EUR
ENZOENZKIT1310001
TSH ELISA kit
Tests Tests ELISA
Highly sensitive TSH ELISA kit enabling detection of TSH in serum in just 75 minutes.
  • Sensitive measurement of TSH, detecting as little as 0,5 µIU/ml
  • Negligible reactivity from similar glycoprotein hormones
  • High throughput format with results in 75 minutes
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of TSH in serum in human samples.
The TSH ELISA kit is a colorimetric immunometric immunoassay kit with results in 75 minutes.
Informations de commande: Contains Microtiter Plate, Standard, Wash Buffer Concentrate, Conjugate, Substrate, and Stop Solution.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR