Imprimer
CRP (Human) ELISA kit, Grow Cells
Fournisseur: ENZO LIFE SCIENCES
ENZKIT1020001
ENZOENZKIT1020001EA 438 EUR
ENZOENZKIT1020001
CRP (Human) ELISA kit, Grow Cells
Tests Tests ELISA
Sensitive (12,685ng/ml) ELISA kit for the quantification of CRP, an early biomarker for inflammation
  • Sensitive measurement of CRP, detecting as little as 12,685 ng/ml
  • Negligible reactivity for similar family members
  • High throughput format with results in <2 hours for up to 40 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of human C-reactive protein in serum and plasma.
The CRP (human) ELISA kit is a complete, colorimetric, competitive immunoassay kit for the quantitative determination of human CRP in serum and plasma with results in less than two hours.
Informations de commande: Contains Microtiter Plate, Assay Buffer, Standard, Conjugate, Antibody, Streptavidin-HRP, TMB Substrate, Stop Solution, Wash Buffer Concentrate.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR