Imprimer
EpiXtract® Nuclear protein isolation kit II (nucleic acid-free)
Fournisseur: ENZO LIFE SCIENCES
ENZ450150100
ENZOENZ450150100EA 299 EUR
ENZOENZ450150100
EpiXtract® Nuclear protein isolation kit II (nucleic acid-free)
Protéines extraction Protein Extraction Kits
Rapid, reliable method to isolate nucleic acid-free nuclear proteins in mammalian cells and tissues.
For the extraction of nucleic acid-free nuclear proteins from mammalian cells or tissues.
EpiXtract® Nuclear Protein Isolation Kit II provides a simple and reliable method to isolate nucleic acid-free nuclear proteins. Extracts can be used for a variety of downstream applications including western blotting, protein-DNA binding assays, nuclear enzyme assays and others requiring nucleic acid-free nuclear proteins.
Informations de commande: Contains Pre-extraction Buffer, Extraction Buffer, Extraction pre-cleaner, Extraction cleaner, Protease Inhibitor Cocktail, DTT.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR