Imprimer
DNA methylation kit, 5-hmC, BIOARRAY™
Fournisseur: ENZO LIFE SCIENCES
ENZ450110050
ENZOENZ450110050EA 819 EUR
ENZOENZ450110050
DNA methylation kit, 5-hmC, BIOARRAY™
Tests Tests à l'acide nucléique
BIOARRAY™ 5-hmC Methylation Kit is used for the sequence specific detection of 5-hydroxymethylcytosine within DNA using a simple and efficient glucosylation reaction procedure.
  • Single tube reaction format
  • Suitable for analysis by a variety of downstream applications
  • Flexible set-up for custom assay design
Information de livraison: Kit contains 5-hmC Glucosyltransferase (5-hmC), 10X 5-hmC reaction buffer, 10X UDPG (uridine diphosphoglucose), MspI restriction enzyme, 5-hmC control DNA, qPCR primer 1, qPCR primer 2 and 5-DNA clean up concentrator kit.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR