Imprimer

LRRKtide

Fournisseur: ENZO LIFE SCIENCES

BMLP2670001
ENZOBMLP2670001EA 159 EUR
ENZOBMLP2670001
LRRKtide
Protéines et Peptides

LRRK substrate


This peptide, RLGRDKYKTLRQIRQ, is derived from ezrin/moesin/radixin and is a substrate for LRRK kinase.

New Product

Commander maintenant

SPÉCIFICATIONS

Nom de la protéine/du peptide LRRKtide
Environmentally Preferable
Species
Source
Purity >95% (HPLC)
Formulation Lyophilised

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR