Imprimer
Matrix Metalloproteinase-13 (MMP-13) colorimetric drug discovery kit
Fournisseur: ENZO LIFE SCIENCES
BMLAK4120001
ENZOBMLAK4120001EA 632 EUR
ENZOBMLAK4120001
Matrix Metalloproteinase-13 (MMP-13) colorimetric drug discovery kit
Tests Tests enzymatiques
A QuantiZyme™ Assay System. The MMP-13 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-13 (MMP-13, collagenase-3) using a thiopeptide as a chromogenic substrate.
Information de livraison: Kit contains MMP-13 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR