Imprimer

2-Phosphonomethyl pentanedioic acid 97 HPLC_ASSAY_METHOD

Fournisseur: ENZO LIFE SCIENCES

Synonyms: 2-(Phosphonomethyl)pentanedioic acid, 2-PMPA

ALX550358M005
ENZOALX550358M005EA 366 EUR
ENZOALX550358M005
2-Phosphonomethyl pentanedioic acid 97 HPLC_ASSAY_METHOD
2-Phosphonomethyl pentanedioic acid

Potent inhibitor of the neuropeptidase N-acetylated α-linked acidic dipeptidase (NAALADase), which is a membrane-bound peptidase that hydrolyses N-acetyl-L-aspartate-L-glutamate (NAAG), a major brain peptide.


  • NAALADase inhibitor


Viscous, colorless oil.


Soluble in water, methanol or DMSO.

Formule: C₆H₁₁O₇P
MW: 226.12 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD00940167
Numéro CAS: 173039-10-6

Commander maintenant

SPÉCIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR