Imprimer
IFN-gamma (human), ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI900136
ENZOADI900136EA 822 EUR
ENZOADI900136
IFN-gamma (human), ELISA kit
Tests Tests ELISA
The IFN-gamma (Human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450 nm.
  • Ultrasensitive
  • Efficient - only 50 µL diluted sample needed per well
  • Measure natural and recombinant human IFN-γ
For the quantitative determination of human IFN-gamma in culture supernatants, plasma, serum, and urine.
Information de livraison: Supplied with microtiter plate, SA-HRP concentrate, SA-HRP dilution buffer, biotinylated antibody, standard, standard diluent, wash buffer concentrate, TMB substrate, stop solution
Plus About VWR