Imprimer
TNF alpha (mouse) ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI900047
ENZOADI900047EA 695 EUR
ENZOADI900047
TNF alpha (mouse) ELISA kit
Tests Tests ELISA
For the quantitative determination of mouse TNF-α in culture supernatants and serum. Cited sample type includes tissue and plasma.
The TNF-α (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 5 hours. Absorbance is read at 450 nm.
Informations de commande: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 16, Wash buffer concentrate, Standard, TMB Substrate, Stop solution.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR