Imprimer
Tank, rectangular, PFA
Fournisseur: ENTEGRIS
E92-01D-0215
ENTEE92-01D-0215EA 0 EUR
ENTEE92-01D-0215
Tank, rectangular, PFA
Récipients Récipients rectangulaires
Injection molded rectangular tanks with dual capacity.
  • Available in single or dual capacity
  • Available with or without covers
  • Tanks should be properly supported in heated applications
Maximum recommended working temperature: 180 °C.
Mounting hole dimension (L×W): 419×216 mm.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR