Imprimer
Forceps, PFA
Fournisseur: ENTEGRIS
C20-0215
ENTEC20-0215EA 0 EUR
ENTEC20-0215
Forceps, PFA
Pinces
PFA, white.
  • Straight forceps for substrate handling
  • Extra heavy for much stronger grip at ends
Conditionnement: Packaged in protective cases.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR