Imprimer
Measuring Cables, CYK10
Fournisseur: Endress+Hauser
CYK10-A031 CYK10-A051 CYK10-A101
ENDRCYK10-A031EA 150 EUR
ENDRCYK10-A031 ENDRCYK10-A051 ENDRCYK10-A101
Measuring Cables, CYK10
Câbles
CYK10 measuring cable is used with all sensors with Memosens plug-in head. Forget about moisture and benefit from completely safe signal transmission.
  • Memosens data cable for all sensors with Memosens plug-in head
  • Maximum process safety via non-contact, inductive and digital signal transmission
  • No contact corrosion, absolutely watertight, no interference potential
  • Increased availability of the measuring point: Digital data transmission provides an automatic error message if the signal is lost
CYK10 guarantees absolute water-proof connection ofMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR