Imprimer
Pipette tracker, LightOne™ Illuminator FS, Embitec®
Fournisseur: EMBITEC
LI-1200
EMBILI-1200EA 745 EUR
EMBILI-1200
Pipette tracker, LightOne™ Illuminator FS, Embitec®
Transilluminateurs Transilluminateurs LED
Compact benchtop illuminator that helps visually identify a specific sample well, complete lanes or rows, to aid in loading 96-well plates.
  • White LED's which have three distinct intensities
  • Auto and manual/hands-free advance modes
  • Dedicated LED for each sample well to provide accurate guidance
  • User defined time intervals for advancing to the next sample(s)
  • Illuminate single well or complete lane or complete row
  • Material: White polycarbonate
The LightOne™ Illuminator FS is a stand-alone bench top solution for tracking pipetting with enhancedMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR