Imprimer
Cole-Parmer® BB-200 Electric Bunsen Burners
Fournisseur: Cole-Parmer
36135-02 36135-01
ELET36135-02EA 1340 EUR
ELET36135-02 ELET36135-01
Cole-Parmer® BB-200 Electric Bunsen Burners
Appareils chauffants Brûleurs à gaz
Provide clean and easy operation.
  • Top cowl deflects heat away from hands
  • Air circulation from vented housing keeps base cool to touch
  • Energy efficient - only consume 400 W of power
These electric Bunsen burners combine the advantages of a regular gas burner with the clean and easy operation of a heating mantle. Corrosion-resistant stainless steel burners features a cone-shaped coiled resistance heater which directs radiant heat out from the cavity. Cooling vents circulate air to cool the base, making it possible to hold while in use. A detachable cowlMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR