Imprimer
Rotary Vane Vacuum Pump
Fournisseur: EDWARDS HIGH VACUUM METTLER
A34315903 A37310940 A37317984 A37330940
EDWAA34315903EA 0 EUR
EDWAA34315903 EDWAA37310940 EDWAA37317984 EDWAA37330940
Rotary Vane Vacuum Pump
Pompe Pompes à vide à palettes
The E1M18 and E2M18 pumps are direct drive, sliding vane pumps. The E1M18 is a single-stage pump, and the E2M18 is a two-stage pump. The pump is oil sealed and designed for reliable, long-term operation in both laboratory and industrial environments. The pump is a freestanding unit with the drive provided through a flexible coupling by either a single-phase or three-phase (four-pole) motor.
  • Easy to use controls
  • Air tight pumping chambers prevents oil suck back unless the gas ballast valve is open
  • An oil distribution valve which prevents discharge of oil to
More Product Information
Plus About VWR