Imprimer
EBI 12-T220 Temperature Data Loggers for Process Validation
Fournisseur: EBRO
1340-6604 1340-6605
EBRO1340-6604EA 4120 EUR
EBRO1340-6604 EBRO1340-6605
EBI 12-T220 Temperature Data Loggers for Process Validation
Enregistreurs de données
The EBI 12-T220 is a precise temperature data logger for process validation in laboratories and the pharmaceutical industry. With a temperature range of –200 to +200 °C and an accuracy of up to 0,05 °C, it enables precise temperature measurements in critical applications such as sterilisation and disinfection.
  • Suitable for steam sterilisers, washer–disinfectors
  • Temperature range –200 to +200 °C
  • High accuracy up to 0,05 °C
  • Bendable metal probe (Ø 1,50 mm)
The EBI 12-T220 temperature data logger from ebro fulfils the stringent requirements of the
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR