Imprimer
Safety Cabinets for Storage of Flammable Liquids, Type 90, COMPACT line
Fournisseur: DUPERTHAL
29-201777-011 29-200977-031
DUEH29-201777-011EA 6228.36 EUR
DUEH29-201777-011 DUEH29-200977-031
Safety Cabinets for Storage of Flammable Liquids, Type 90, COMPACT line
Mobilier Armoires de stockage
Classical door technology (door handle) for the storage of flammable liquids type testing by MPA/TÜV SÜD according to DIN EN 14470-1, DIN EN 16121 and DIN EN 16122, EK5/AK4 09-10:2009 and AfPS GS 2014: 01 PAK mark of approval: GS/CE mark, high quality mark of approval by TÜV SÜD construction and colour.
  • Outer carcass sheet steel, RAL 7035
  • Doors sheet steel, RAL 1018
  • Inner carcass of decor panels, RAL 7035
  • Doors close automatically in case of fire
  • Doors remain open in every position
  • Cylinder lock for safe access
  • Bottom tray sheet steel, RAL 7035
  • Hot-dip
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR