Imprimer
Test sets
Fournisseur: Dräger
6400090 8101735 8101736 8101770 8103170 8103380 8103410
151-0116EA 79.9 EUR
151-0116 151-0176 151-0177 151-0179 DRAE8103170 DRAE8103380 DRAE8103410
Test sets
Tubes de détection de gaz
These test sets allow for the semi quantitative measurements of five different gases at the same time.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR