Imprimer
Supelco® TraceCERT® Organic Reference Standard, Aldicarb-sulfone
Fournisseur: Merck
CRM69033-100MG
CRM69033-100MGEA 151 EUR
CRM69033-100MG
Supelco® TraceCERT® Organic Reference Standard, Aldicarb-sulfone
Étalons Étalons organiques
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Attention: P260 - P262 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR