Imprimer
Twin Bucket
Fournisseur: CONTEC
2743S
CONT2743SEA 435 EUR
CONT2743S
Twin Bucket
Godets
Twin Bucket is an autoclavable 20 L divided bucket made of durable PP with a curved stainless steel wringer.
  • Ideal for smaller controlled environments and labs with limited storage space
  • Designed to work with the curved stainless steel wringer
  • Four castors for added mobility
  • Fully autoclavable (except castors)
One side of the bucket can be used for fresh cleaning and disinfecting solutions, and the other one for waste.
Information de livraison: Supplied with lid, stainless steel wringer and four castors.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR