Imprimer
Safety helmets, Vision Plus
Fournisseur: CENTURION SAFETY PRODUCTS
S10PLUSEWR
CENUS10PLUSEWREA 52.15 EUR
CENUS10PLUSEWR
Safety helmets, Vision Plus
Casques Casques de sécurité
The Vision Plus helmet offers wearers of prescription glasses total safety reassurance – with moulded integrated eye protection certified to grade A high impact levels. Helmet shell is made from ABS plastic.
  • Optical clarity to Class 1 for continuous use
  • Large side pull tabs enable the wearer to quickly and easily move the visor up and down even when wearing gloves
  • Moulded design provides improved lower cheek coverage
  • Anti-scratch and anti-mist coating for improved durability and visibility
  • Wheel ratchet, non-vented
Weight: 525 g with fitted visor
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR